L-Lysine methyl ester dihydrochloride 98%

Synonyms: H-Lys-OMe.2HCl

AC45927-0250
103518-612EA 61.1 USD
103518-612
L-Lysine methyl ester dihydrochloride 98%
L-Lysine methyl ester dihydrochloride
Formula: C₇H₁₆N₂O₂
MW: 233.13 g/mol
Melting Pt: 200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039067
CAS Number: 26348-70-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Specific optical rotation +21° to +25° (20 °C, 589 nm) (1% in MeOH)
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR