Lithium sulfate, anhydrous for analysis

Warning

AC44755-2500
103516-906EA 76.07 USD
103516-906
Lithium sulfate, anhydrous for analysis
Lithium sulphate
Formula: Li₂O₄S
MW: 109.95 g/mol
Melting Pt: 859 °C
Density: 2.22 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011086
CAS Number: 10377-48-7

Order Now

Specification Test Results

Appearance White Crystalline powder
Water ≤1 %
Solubility (10 % in water) Clear colorless to slightly hazy
Titration after Ion exchange ≥98.5 %
Calcium (Ca) ≤0.02 %
Iron (Fe) ≤0.005 %
Magnesium (Mg) ≤0.02 %
Potassium (K) ≤0.02 %
Heavy metals (as Pb) ≤0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR