Trilithium orthophosphate, Extra Pure

Warning

Synonyms: Lithium orthophosphateLithium phosphateLithium phosphate tribasic

AC38978-1000
103513-092EA 42.6 USD
103513-092
Trilithium orthophosphate, Extra Pure
tri-Lithium orthophosphate
Formula: Li₃O₄P
MW: 115.79 g/mol
Melting Pt: 837 °C
Storage Temperature: Ambient
MDL Number: MFCD00016187
CAS Number: 10377-52-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration with HClO4 ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR