Lithium nitrate 99.999% (trace metals basis)

Warning

AC21254-1000 AC21254-0050 AC21254-0250
103524-174EA 572 USD
103524-174 103524-170 103524-172
Lithium nitrate 99.999% (trace metals basis)
Lithium nitrate
Formula: LiNO₃
MW: 68.95 g/mol
Boiling Pt: 600 °C (760 mmHg)
Melting Pt: 251 °C
Density: 2.38 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011094
CAS Number: 7790-69-4
UN: 2722
ADR: 5.1,III

Order Now

Specification Test Results

Appearance White Crystalline powder and/or chunks
Total trace metal impurities ≤10 ppm
Lithium (Li) 9 to 11 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR