Lithiumbis(trimethylsilyl)amide 1 M in THF, AcroSeal™

Danger

AC20955-1000 AC20955-8000
103526-656EA 51 USD
103526-656 103526-658
Lithiumbis(trimethylsilyl)amide 1 M in THF, AcroSeal™
Lithium bis(trimethylsilyl)amide
Formula: C₆H₁₈LiNSi₂
MW: 167.33 g/mol
Boiling Pt: 65 °C (760 mmHg)
Density: 0.9 g/cm³
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD00008261
CAS Number: 4039-32-1
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Light yellow to amber Liquid
Strength 0.90 to 1.10 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR