Aspartame

Supplier: Thermo Scientific
AC22865-0010 AC22865-0050
103509-904EA 160 USD
103509-904 103511-744
Aspartame
Proteins and Peptides
  • Infrared spectrum: Conforms
  • Loss on drying: ≤4.5% (105 °C, 4 hrs)
  • UV transmittance: ≥95% (c=1, 2 N HCl, 430 nm) 1 cm cell
  • Specific optical rotation: +12.5 to +17.5° (20 °C, 589 nm) (c=4, 15 N formic acid) (within 30 min)
  • Heavy metals (as Pb): ≤10 ppm
  • Sulfated ash: ≤0.2%
  • Melting point: 248 to 250 °C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR