Inulin (from chicory root)

Synonyms: Inulin (by Enzymatic synthesis)DahlinAlantin

AC45710-0250 AC45710-5000
103518-458EA 93.1 USD
103518-458 103518-462
Inulin (from chicory root)
Inulin
Storage Temperature: Ambient
MDL Number: MFCD00131407
CAS Number: 9005-80-5

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Loss on drying ≤10 % (2 g, 105 °C, 2 hrs)
Free fructose, glucose,sucrose ≤5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR