Imidazo[1,2-a]pyridine-2-carboxylic acid hydrate 97%

Warning

AC43278-0050 AC43278-0010
103515-460EA 483 USD
103515-460 103515-458
Imidazo[1,2-a]pyridine-2-carboxylic acid hydrate 97%
Imidazo[1,2-a]pyridine-2-carboxylic acid
Formula: C₈H₆N₂O₂·xH₂O
Melting Pt: 232…234 °C
Storage Temperature: Ambient
MDL Number: MFCD17171427
CAS Number: 64951-08-2

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Additional info Hydrate, contains 9.5% to 11.0% water
HPLC ≥96.0 %
Elemental Analysis 51.20 to 55.47 % C
Elemental Analysis 4.30 to 4.65 % H
Elemental Analysis 14.96 to 16.17 % N
Residue after evaporation ≤0.01 %
Peroxides ≤50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR