1-Hexanol, anhydrous 99%, AcroSeal™
|
Warning
|
Synonyms:
Hexyl alcohol
, 1-Hydroxy hexane
, 1-Hexyl alcohol
, Pentyl carbinol
AC43386-0010
AC43386-1000
103528-842EA
35.6
USD
103528-842
103528-840
1-Hexanol, anhydrous 99%, AcroSeal™
1-Hexanol
Formula:
H₃C(CH₂)₄CH₂OH MW: 102.18 g/mol Boiling Pt: 156…157 °C (760 mmHg) Melting Pt: –52 °C Density: 0.8136 g/cm³ Flash Pt: 59 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002982 CAS Number: 111-27-3 UN: 2282 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.005 % |
GC | ≥98.5 % |
Residue after evaporation | ≤0.002 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...