7-Isopropyl-1,4-dimethylazulene 99%
|
Warning
|
Synonyms:
Guaiazulene
AC12020-0250
AC12020-0100
103509-496EA
53.6
USD
103509-496
103509-498
7-Isopropyl-1,4-dimethylazulene 99%
7-Isopropyl-1,4-dimethylazulene
Formula:
C₁₅H₁₈ MW: 198.31 g/mol Boiling Pt: 153 °C (7Torr) Melting Pt: 29…31 °C Density: 0.97 g/cm³ Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003811 CAS Number: 489-84-9 EINECS: 207-701-2 |
Specification Test Results
Appearance | Dark blue Low melting crystalline solid |
Infrared spectrum | Conforms |
GC | ≥98.5 % |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...