Gold(I) chloride 99+%

Danger

AC38833-0050 AC38833-0010
103528-306EA 785 USD
103528-306 103528-304
Gold(I) chloride 99+%
Gold (I) chloride
Formula: AuCl
MW: 232.42 g/mol
Melting Pt: 289 °C
Storage Temperature: Ambient
MDL Number: MFCD00046175
CAS Number: 10294-29-8
EINECS: 233-655-8
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Appearance Yellow-green Powder
Elemental Analysis ≥99.0 % (ex Au)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR