Gallic acid monohydrate ACS

Warning

Synonyms: 3,4,5-Trihydroxybenzoic acid hydrate3,4,5-Trihydroxybenzoic acid monohydrate

AC44758-5000 AC44758-1000
103516-916EA 200.77 USD
103516-916 103516-908
Gallic acid monohydrate ACS
Gallic acid monohydrate
Formula: C₇H₆O₅·1H₂O
MW: 188.14 g/mol
Melting Pt: 251 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149098
CAS Number: 5995-86-8

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Residue after ignition ≤0.05 %
Titration with NaOH ≥98.0 %
Sulfate (SO4) ≤0.02
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR