Fumonisin B2 96+%

Danger

Synonyms: 1,2,3-Propanetricarboxylic acid, 1,1'-(1-(2-amino-9,11-dihydroxy-2-methyltridecyl)-2-(1-methylpentyl)-1,2-ethanediyl) ester1,2,3-Propanetricarboxylic acid, 1,1?-[1-(12-amino-4,11-dihydroxy-2-methyl-tridecyl)-2-(1-methylpentyl)-1,2-ethane-diyl]esterFumonisin B2 from Fusarium moniliforme, sphingosine N-acyltransferase inhibitor

AC32826-0010
103524-242EA 377 USD
103524-242
Fumonisin B2 96+%
Fumonisin B2
Formula: C₃₄H₅₉NO₁₄
MW: 705.839 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00133350
CAS Number: 116355-84-1
UN: 2811
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR