2-Fluoro-α-methyl-4-biphenylacetic acid 98%

Danger

Synonyms: 2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acidFrobenAnsaidAntadysFlurbiprofen

AC45466-0010 AC45466-0050 AC45466-0250
103529-046EA 236 USD
103529-046 103529-042 103529-044
2-Fluoro-α-methyl-4-biphenylacetic acid 98%
2-Fluoro-α-methyl-4-biphenylacetic acid
Formula: C₁₅H₁₃FO₂
MW: 244.265 g/mol
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00079303
CAS Number: 5104-49-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 110 to 116 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR