5-Aminofluorescein (isomer I), pure

Warning

Synonyms: 5-Amino-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-oneFluoresceinamine, isomer I5-Amino-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

AC40077-0050
103514-644EA 258 USD
103514-644
5-Aminofluorescein (isomer I), pure
5-Aminofluorescein (isomer I)
Formula: C₂₀H₁₃NO₅
MW: 347.33 g/mol
Melting Pt: 223 °C
Storage Temperature: Ambient
MDL Number: MFCD00005052
CAS Number: 3326-34-9

Order Now

Specification Test Results

Appearance Dark red-brown Crystalline powder
Infrared spectrum Conforms
HPLC 95 % Typical
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR