Fluconazole 98%

Warning

Synonyms: 2-(2,4-Difluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol2-(2,4-Difluorophenyl)-1,3-di(1H-1,2,4-triazol-1-yl)propan-2-ol

AC45548-0010 AC45548-0050
103517-618EA 138.32 USD
103517-618 103517-620
Fluconazole 98%
Fluconazole
Formula: C₁₃H₁₂F₂N₆O
MW: 306.28 g/mol
Melting Pt: 137…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00274549
CAS Number: 86386-73-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 138 to 142 °C
Loss on drying ≤1 % (105 °C, 3 h)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR