2-Ethyl-3-hydroxy-4-pyrone 98%

Warning

Synonyms: 2-Ethyl-3-hydroxy-4H-pyran-4-oneEthyl Maltol

AC45080-1000 AC45080-0250
103517-106EA 77.5 USD
103517-106 103517-104
2-Ethyl-3-hydroxy-4-pyrone 98%
2-Ethyl-3-hydroxy-4-pyrone
Formula: C₇H₈O₃
MW: 140.14 g/mol
Boiling Pt: 289…290 °C
Melting Pt: 89…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00059795
CAS Number: 4940-11-8

Order Now

Specification Test Results

Appearance White to cream Crystalline powder or solid
Infrared spectrum Conforms
Melting point 88 to 94 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR