Ethyl isodehydroacetate 95%

Synonyms: Ethyl 4,6-dimethyl-2-oxo-2H-pyran-5-carboxylate

AC37390-0250 AC37390-0050
103512-950EA 30.6 USD
103512-950 103512-952
Ethyl isodehydroacetate 95%
Ethyl isodehydroacetate
Formula: C₁₀H₁₂O₄
MW: 196.1999 g/mol
Boiling Pt: 290…295 °C (760 mmHg)
Melting Pt: 17…18 °C
Density: 1.167 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006643
CAS Number: 3385-34-0
EINECS: 222-195-3

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5135 to 1.5165 (20 °C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR