Ethyl homovanillate 97%

Warning

Synonyms: 4-HYDROXY-3-METHOXYPHENYLACETIC acid ETHYL ESTEREthyl 2-(4-hydroxy-3-methoxyphenyl)acetate

AC18924-0010
103511-130EA 48.76 USD
103511-130
Ethyl homovanillate 97%
Ethyl homovanillate
Formula: C₁₁H₁₄O₄
MW: 210.23 g/mol
Boiling Pt: 180…185 °C (14 mmHg)
Melting Pt: 44…47 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00009180
CAS Number: 60563-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR