Ethyl-6-chloronicotinate 97%

Warning

Synonyms: Ethyl 6-chloronicotinate

AC45464-1000 AC45464-0050 AC45464-0250
103517-556EA 289.4 USD
103517-556 103517-554 103517-552
Ethyl-6-chloronicotinate 97%
Ethyl 6-chloronicotinate
Formula: C₈H₈ClNO₂
MW: 185,61 g/mol
Boiling Pt: 98 °C (1.5 mmHg)
Melting Pt: 26…30 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00082739
CAS Number: 49608-01-7

Order Now

Specification Test Results

Appearance Colorless to white to yellow Low melting solid or liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR