Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate 98%

Warning

Synonyms: Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylateEthyl 4-chloro-2-methylthio-5-pyrimidinecarboxylateEthyl 4-chloro-2-(methylsulfanyl)pyrimidine-5-carboxylateEthyl 4-chloro-2-(methylsulphanyl)pyrimidine-5-carboxylate

AC16375-0050
103510-812EA 44.7 USD
103510-812
Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate 98%
Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate
Formula: C₈H₉ClN₂O₂S
MW: 232.69 g/mol
Melting Pt: 60…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00006085
CAS Number: 5909-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR