Ethyl-2-methylnicotinate 97%

Warning

Synonyms: Ethyl 2-methylnicotinate

AC43619-0010 AC43619-0050
103515-774EA 61.2 USD
103515-774 103515-772
Ethyl-2-methylnicotinate 97%
Ethyl 2-methylnicotinate
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 126…127 °C (24 mmHg)
Density: 1.07 g/cm³
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00006336
CAS Number: 1721-26-2

Order Now

Specification Test Results

Appearance Clear colorless to orange Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR