Ethyl-2-cyanoacrylate
|
Warning
|
Synonyms:
Ethyl 2-cyanoacrylate
AC43445-0050
103524-420EA
101
USD
103524-420
Ethyl-2-cyanoacrylate
Ethyl-2-cyanoacrylate
Formula:
C₆H₇NO₂ MW: 125.1253 g/mol Boiling Pt: 54…56 °C (760 mmHg) Melting Pt: –20 °C Density: 1.04 g/cm³ Flash Pt: 85 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00045615 CAS Number: 7085-85-0 UN: 3335 ADR: 9,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
NMR | Consistent with structure |
NMR (1H-NMR) | Consistent with structure |
Titration with HClO4 | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...