Ethyl-2-amino-4,5-dimethylthiophene-3-carboxylate 97%

Warning

Synonyms: Ethyl 2-amino-4,5-dimethylthiophene-3-carboxylateEthyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate

AC45030-0010 AC45030-0100
103517-204EA 227.9 USD
103517-204 103517-202
Ethyl-2-amino-4,5-dimethylthiophene-3-carboxylate 97%
Ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate
Formula: C₉H₁₃NO₂S
MW: 199.27 g/mol
Melting Pt: 90…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00085051
CAS Number: 4815-24-1

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline powder
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR