Ethanethiol 99+%

Danger

Synonyms: 1-MercaptoethaneEthyl mercaptan

AC11786-5000 AC11786-0025 AC11786-0500
103525-034EA 21 USD
103525-034 103525-036 103525-032
Ethanethiol 99+%
Ethanethiol
Formula: C₂H₆S
MW: 62.14 g/mol
Boiling Pt: 35 °C (760 mmHg)
Melting Pt: –148 °C
Density: 0.83 g/cm³
Flash Pt: –45 °C
Storage Temperature: Ambient
MDL Number: MFCD00004887
CAS Number: 75-08-1
EINECS: 200-837-3
UN: 2363
ADR: 3,I

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4296 to 1.4316 (20 °C, 589 nm)
GC ≥99.0 %
Cloud point ≤20.0 °C
Calcium (Ca) ≤30 ppm
Iron (Fe) ≤10 ppm
Sodium (Na) ≤200 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR