Estriol 97%

Danger

AC46049-0010 AC46049-0050
103518-640EA 107 USD
103518-640 103518-642
Estriol 97%
Estriol
Formula: C₁₈H₂₄O₃
Melting Pt: 280…282 °C
Storage Temperature: Ambient
MDL Number: MFCD00003691
CAS Number: 50-27-1

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥96.0 %
Specific optical rotation +54° to +62° (20 °C, 589 nm) (0.4 % in Dioxane)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR