D-Psicose 98%

Synonyms: (3R,4R,5R)-1,3,4,5,6-Pentahydroxyhexan-2-one

AC45627-0050
103517-494EA 1476 USD
103517-494
D-Psicose 98%
D-Psicose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00083478
CAS Number: 551-68-8

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Water ≤25 %
HPLC ≥97.5 % (ELSD)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR