DL-Isoserine 98%

Synonyms: DL-3-Amino-2-hydroxypropionic Acid

AC27967-2500
103509-968EA 61.2 USD
103509-968
DL-Isoserine 98%
DL-Isoserine
Formula: C₃H₇NO₃
MW: 105.09 g/mol
Melting Pt: 235 °C
Storage Temperature: Ambient
MDL Number: MFCD00008138
CAS Number: 565-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR