(±)-2-Aminobutyric acid 99%

Danger

Synonyms: H-DL-Abu-OHDL-2-Aminobutyric acidDL-α-Amino-N-Butyric Acid

AC17209-1000 AC17209-0250
103510-712EA 46 USD
103510-712 103510-714
(±)-2-Aminobutyric acid 99%
(±)-2-Aminobutyric acid
Formula: C₄H₉NO₂
MW: 103.12 g/mol
Melting Pt: 291…293 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008093
CAS Number: 2835-81-6
EINECS: 220-616-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR