DL-α-Aminoadipic acid, anhydrous 99%

Synonyms: 2-Aminohexanedioic acidH-DL-Aad-OHH-DL-Homoglu-OHDL-2-Aminoadipic acid(RS)-2-Aminohexanedioic acid

AC44179-0010 AC44179-0050
103516-174EA 73.1 USD
103516-174 103516-176
DL-α-Aminoadipic acid, anhydrous 99%
DL-α-Aminoadipic acid
Formula: C₆H₁₁NO₄
MW: 161.16 g/mol
Melting Pt: 176…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00063119
CAS Number: 542-32-5

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with NaOH 98.5 to 101.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR