Di-tert-butyl phosphonate 96%

Warning

Synonyms: Di-tert-butyl phosphite

AC44939-0010 AC44939-0250 AC44939-0050
103517-118EA 200 USD
103517-118 103517-116 103517-114
Di-tert-butyl phosphonate 96%
Di-tert-butyl phosphonate

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4210 (20 °C, 589 nm)
GC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR