Di-tert-butyl peroxide 99%

Danger

Synonyms: tert-Butyl peroxideLuperox(R) DI, tert-Butyl peroxide

AC34993-1000 AC34993-0050 AC34993-2500
103527-774EA 29 USD
103527-774 103527-776 103527-778
Di-tert-butyl peroxide 99%
Di-tert-butyl peroxide
Formula: C₈H₁₈O₂
MW: 146.23 g/mol
Boiling Pt: 109…110 °C
Melting Pt: –30 °C
Density: 0.8 g/cm³
Flash Pt: 6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008803
CAS Number: 110-05-4
EINECS: 203-733-6
UN: 3107
ADR: 5.2,

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3880 to 1.3900 (20 °C, 589 nm)
GC ≥98.5 %
GC ≤0.1 % Tert-butyl hydroperoxide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR