Diphenyl-2-pyridylphosphine 97%

Warning

Synonyms: 2-(Diphenylphosphino)pyridine

AC42766-0050
103514-992EA 111 USD
103514-992
Diphenyl-2-pyridylphosphine 97%
Diphenyl-2-pyridylphosphine
Formula: C₁₇H₁₄NP
MW: 263.28 g/mol
Melting Pt: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00192108
CAS Number: 37943-90-1

Order Now

Specification Test Results

Appearance White to yellow to orange Crystalline powder
Infrared spectrum Conforms
Melting point 81 to 86 °C
Elemental Analysis 74.85 to 80.27 % C
Elemental Analysis 5.17 to 5.55 % H
Elemental Analysis 5.1 to 5.6 % N

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR