Dimethyl sulfate 99+%, AcroSeal™

Danger

AC43083-1000
103524-368EA 33.9 USD
103524-368
Dimethyl sulfate 99+%, AcroSeal™
Dimethyl sulphate
Formula: C₂H₆O₄S
MW: 126.13 g/mol
Boiling Pt: 188 °C (760 mmHg)
Melting Pt: –32 °C
Density: 1.33 g/cm³
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00008416
CAS Number: 77-78-1
UN: 1595
ADR: 6.1,I

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Oily liquid
Infrared spectrum Conforms
Refractive index 1.3860 to 1.3880 (20 °C, 589 nm)
Acidity ≤0.2 % (as Sulfuric acid)
Titration 99.0 to 101.0 % (Hydrolysis)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR