Digoxin 95%

Danger

AC22812-0050 AC22812-0010
103527-044EA 426 USD
103527-044 103527-042
Digoxin 95%
Digoxin
Formula: C₄₁H₆₄O₁₄
Melting Pt: 248…250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003674
CAS Number: 20830-75-5
EINECS: 244-068-1
UN: 3462
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤1 % (500 mg) (vacuum)
Sulfated ash ≤0.1 %
HPLC ≥94 % (on dried substance)
Specific optical rotation +10° to +13° (20 °C, 589 nm) (c=2, anhydr. pyridine) (on dried substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR