Diethyl pyrocarbonate 97%, pure

Warning

Synonyms: Diethyl dicarbonateDEPC (Diethylpyrocarbonate)DEPC

AC17025-1000 AC17025-0050 AC17025-5000 AC17025-0250 AC17025-0025
103516-546EA 96.29 USD
103516-546 103516-548 103516-542 103516-544 103516-550
Diethyl pyrocarbonate 97%, pure
Diethyl pyrocarbonate
Formula: O(COOC₂H₅)₂
MW: 162.14 g/mol
Boiling Pt: 93…94 °C (18 mmHg)
Density: 1.1175 g/cm³
Flash Pt: 69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009106
CAS Number: 1609-47-8
EINECS: 216-542-8
Merck Index: 12,08182

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR