Diethyl carbonate, anhydrous ≥99%, AcroSeal™

Supplier: ACROS ORGANICS MS

Warning

AC43613-0010 AC43613-1000
103528-852EA 255.52 USD
103528-852 103528-854
Diethyl carbonate, anhydrous ≥99%, AcroSeal™
Diethyl carbonate
Formula: C₅H₁₀O₃
MW: 118,13 g/mol
Boiling Pt: 126°C - 128°C (760mmHg)
Melting Pt: –43 °C
Density: 0.975
Flash Pt: 33°C
Storage Temperature: Ambient
MDL Number: MFCD00009107
CAS Number: 105-58-8
EINECS: 203-311-1
UN: 2366
ADR: 3,III
Merck Index: 13,03815

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3830 to 1.3850 (20°C, 589 nm)
Water ≤0.1 %
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR