Diethyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate 95%

Warning

Synonyms: Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylateHantzsch EsterDiethyl 1,4-dihydro-3,5-pyridinecarboxylate1,4-Dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic Acid Diethyl Ester

AC39919-0010
103514-568EA 81.9 USD
103514-568
Diethyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate 95%
Diethyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate
Formula: C₁₃H₁₉NO₄
MW: 253.30 g/mol
Melting Pt: 178…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00005951
CAS Number: 1149-23-1

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥94 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR