Dichloro(methyl)phosphine 90% tech.

Danger

Synonyms: Methyldichlorophosphine

AC31024-0500 AC31024-0100
103527-498EA 194 USD
103527-498 103527-500
Dichloro(methyl)phosphine 90% tech.
Dichloro(methyl)phosphine
Formula: CH₃Cl₂P
MW: 116.91 g/mol
Boiling Pt: 81…82 °C (760 mmHg)
Density: 1.3 g/cm³
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00013616
CAS Number: 676-83-5
EINECS: 211-631-8
UN: 2845
ADR: 4.2,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR