Deuterium oxide 100.0 atom % D, AcroSeal™ for NMR spectroscopy

Synonyms: Heavy waterWater-D2

AC18476-1000
103467-320EA 324 USD
103467-320
Deuterium oxide 100.0 atom % D, AcroSeal™ for NMR spectroscopy
Deuterium oxide
Formula: D₂O
MW: 20 g/mol
Boiling Pt: 101.4 °C (760 mmHg)
Melting Pt: 3.8 °C
Density: 1.105 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00044636
CAS Number: 7789-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR