Decahydronaphthalene (mixture of cis and trans isomers) cis- and trans-mixture, anhydrous 99%, AcroSeal™

Danger

Synonyms: DecalinBicyclo[4.4.0]decaneDecahydronaphthalene

AC40617-1000 AC40617-0010
103528-562EA 38.43 USD
103528-562 103528-560
Decahydronaphthalene (mixture of cis and trans isomers) cis- and trans-mixture, anhydrous 99%, AcroSeal™
Decahydronaphthalene (mixture of cis and trans isomers)
Formula: C₁₀H₁₈
MW: 138.25 g/mol
Boiling Pt: 187 °C (760 mmHg)
Melting Pt: –31 °C
Density: 0.89 g/cm³
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00004130
CAS Number: 91-17-8
EINECS: 202-046-9
UN: 1147
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/82/71/2298271.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR