D-Cycloserine 98%

Synonyms: (R)-(+)-CycloserineD-4-Aminoisoxazolidin-3-oneCycloserineD(+)-Cycloserine(R)-4-Aminoisoxazolidin-3-one

AC22848-0050
103511-738EA 229 USD
103511-738
D-Cycloserine 98%
D-Cycloserine
Formula: C₃H₆N₂O₂
MW: 102.09 g/mol
Melting Pt: 147 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00005353
CAS Number: 68-41-7
EINECS: 200-688-4

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Loss on drying ≤1 % (60 °C, 3 hrs) (vacuum)
UV at 285 nm A: ≤0.8
Condensation products (0.04 % in 0.1 N NaOH)
Potency ≥900 µg/mg
Residue after ignition ≤0.5 %
pH 5.5 to 6.5 (10 % solution)
Specific optical rotation +108° to +114° (20 °C, 589 nm) (c=5, 2N NaOH)
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR