(R)-2-Amino-2-phenylacetamide 99%

Warning

Synonyms: D(-)-PhenylglycinamideH-D-Phg-NH₂·HClH-D-Phg-NH2

AC34670-0250 AC34670-0050
103513-908EA 256 USD
103513-908 103513-906
(R)-2-Amino-2-phenylacetamide 99%
(R)-2-Amino-2-phenylacetamide
Formula: C₈H₁₀N₂O
MW: 150.181 g/mol
Melting Pt: 125…129 °C
Storage Temperature: Ambient
MDL Number: MFCD06799064
CAS Number: 6485-67-2

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
GC ≥98.5 %
Specific optical rotation -110° to -120° (20 °C, 589 nm) (c=0.55, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR