Cytarabine 98%

Warning

Synonyms: Cytosine beta-D-ArabinofuranosideCytosine arabinosideArabinocytidine4-Amino-1-beta-D-arabinofuranosyl-2(1H)-pyrimidinone1-β-D-Arabinofuranosylcytosine4-Amino-1-((2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-oneAra-C

AC44956-1000 AC44956-0010
103517-150EA 49.23 USD
103517-150 103517-146
Cytarabine 98%
Cytarabine
Formula: C₉H₁₃N₃O₅
MW: 243.22 g/mol
Melting Pt: 220 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066487
CAS Number: 147-94-4

Order Now

Specification Test Results

Appearance White to Beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation +155° to +162° (20 °C, 589 nm) (c=1, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR