Cyclopropylmagnesium bromide 0.5 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal™

Danger

AC44595-8000
103524-538EA 1008 USD
103524-538
Cyclopropylmagnesium bromide 0.5 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal™
Cyclopropylmagnesium bromide
Formula: C₃H₅BrMg
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00672094
CAS Number: 23719-80-4
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Colorless to yellow to orange Solution
Titration Grignard 0.40 to 0.60 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR