Cyclopentylboronic acid 95%

Warning

AC35906-0010 AC35906-0100
103515-230EA 56.13 USD
103515-230 103515-232
Cyclopentylboronic acid 95%
Cyclopentylboronic acid
Formula: C₅H₁₁BO₂
MW: 113.95 g/mol
Melting Pt: 134…142 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01074541
CAS Number: 63076-51-7

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or flakes
Infrared spectrum Conforms
Titration with NaOH 94.0 to 106.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR