Cloxacillin sodium salt monohydrate

Danger

AC45530-0050 AC45530-0010
103517-598EA 152 USD
103517-598 103517-596
Cloxacillin sodium salt monohydrate
Cloxacillin sodium salt monohydrate
Formula: C₁₉H₁₈ClN₃O₅S
Storage Temperature: Refrigerator
MDL Number: MFCD00150735
CAS Number: 7081-44-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water 3.0 to 5.0 %
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR