Triammonium citrate 97+%, pure
|
Warning
|
AC39141-0010
AC39141-0500
AC39141-2500
103517-864EA
80.3
USD
103517-864
103518-808
103517-862
Triammonium citrate 97+%, pure
tri-Ammonium citrate
Formula:
C₆H₁₇N₃O₇ MW: 243.22 g/mol Melting Pt: 185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036406 CAS Number: 3458-72-8 |
Specification Test Results
Appearance | White to off-white Crystals |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...