Diammonium hydrogen citrate ≥99% for biochemistry

Supplier: ACROS ORGANICS MS

Warning

Synonyms: Ammonium citrate dibasicDiammonium hydrogen 2-hydroxypropane-1,2,3-tricarboxylateCitric acid diammonium salt

AC44802-5000 AC44802-1000
103518-150EA 156.31 USD InStock
103518-150 103518-148
Diammonium hydrogen citrate ≥99% for biochemistry
di-Ammonium hydrogen citrate
Formula: HOC(CO₂H)(CH₂CO₂NH₄)₂
MW: 226,19 g/mol
Density: 1.5
Storage Temperature: Ambient
MDL Number: MFCD00013068
CAS Number: 3012-65-5
EINECS: 221-146-3
Merck Index: 13,00513

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Solubility Clear colorless
UV (1 M in water)
UV at 260 nm A: ≤0.1
UV at 280 nm A: ≤0.1
Titration with HClO4 ≥99.0 %
Chloride (Cl) ≤0.001 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR