Citicoline 98%

Synonyms: Cytidine 5'-diphosphocholine

AC45778-0050 AC45778-0250 AC45778-0010
103518-490EA 64.61 USD
103518-490 103518-492 103518-494
Citicoline 98%
Citicoline
Formula: C₁₄H₂₆N₄O₁₁P₂
MW: 488.33 g/mol
Melting Pt: 240…242 °C
Storage Temperature: Ambient
MDL Number: MFCD00868097
CAS Number: 987-78-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR